This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NME7 SIRNA Assays ›
Gene Symbol:
NME7
Gene Name:
NME/NM23 family member 7
Gene Aliases:
CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
Chromosome Location:
Chr.1: 169132385 - 169367963 on Build GRCh38
UniGene ID:
Hs.706952
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 45196

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NME7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013330.4 NP_037462.1 2 336
NM_197972.2 NP_932076.1 2 305
NR_104229.1 2 336
XM_017001099.1 XP_016856588.1 2 322
GenBank mRNA AB209049.1 - 2 145
AF153191.2 - 2 175
AK094513.1 - 2 288
AK290701.1 - 2 170
AK301917.1 - 2 319
BC006983.1 - 2 167 3997152
CD107958.1 - 2 364

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
nucleoside diphosphate kinase activity
protein binding
ATP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header