This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SORBS2 SIRNA Assays ›
Gene Symbol:
SORBS2
Gene Name:
sorbin and SH3 domain containing 2
Gene Aliases:
ARGBP2, PRO0618
Chromosome Location:
Chr.4: 185585444 - 185956716 on Build GRCh38
UniGene ID:
Hs.481342
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 44375

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SORBS2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145670.1 NP_001139142.1 8 1254
NM_001145671.2 NP_001139143.1 7 782
NM_001145672.1 NP_001139144.1 6 579
NM_001145673.1 NP_001139145.1 7 1306
NM_001270771.1 NP_001257700.1 9 1263
NM_003603.6 NP_003594.3 7 1096
NM_021069.4 NP_066547.1 6 1043
XM_005263307.1 XP_005263364.1 9 1900
XM_005263308.1 XP_005263365.1 8 1258
XM_005263310.1 XP_005263367.1 8 1189
XM_005263311.1 XP_005263368.1 8 1258
XM_005263312.1 XP_005263369.1 7 1096
XM_005263313.1 XP_005263370.1 8 1258
XM_006714388.1 XP_006714451.1 8 1189
XM_006714390.1 XP_006714453.1 7 1165
XM_017008738.1 XP_016864227.1 10 2374
XM_017008739.1 XP_016864228.1 11 2677
XM_017008740.1 XP_016864229.1 10 2370
XM_017008741.1 XP_016864230.1 10 2374
XM_017008742.1 XP_016864231.1 9 2324
XM_017008743.1 XP_016864232.1 10 2305
XM_017008744.1 XP_016864233.1 10 2584
XM_017008745.1 XP_016864234.1 9 2524
XM_017008746.1 XP_016864235.1 10 2374
XM_017008747.1 XP_016864236.1 10 2515
XM_017008748.1 XP_016864237.1 10 2374
XM_017008749.1 XP_016864238.1 10 1936
XM_017008750.1 XP_016864239.1 10 2374
XM_017008751.1 XP_016864240.1 10 2374
XM_017008752.1 XP_016864241.1 10 2374
XM_017008753.1 XP_016864242.1 10 2374
XM_017008754.1 XP_016864243.1 10 2374
XM_017008755.1 XP_016864244.1 7 1339
XM_017008756.1 XP_016864245.1 6 1042
XM_017008758.1 XP_016864247.1 7 1208
XM_017008759.1 XP_016864248.1 7 1935
XM_017008760.1 XP_016864249.1 7 1165
XM_017008761.1 XP_016864250.1 10 2374
XM_017008762.1 XP_016864251.1 8 1258
XM_017008763.1 XP_016864252.1 7 1208
XM_017008764.1 XP_016864253.1 8 1189
XM_017008765.1 XP_016864254.1 9 1492
XM_017008766.1 XP_016864255.1 7 1165
XM_017008767.1 XP_016864256.1 8 1468
XM_017008768.1 XP_016864257.1 7 1386
XM_017008769.1 XP_016864258.1 8 1189
XM_017008770.1 XP_016864259.1 7 1165
XM_017008771.1 XP_016864260.1 6 1046
XM_017008772.1 XP_016864261.1 7 1096
XM_017008773.1 XP_016864262.1 8 1399
XM_017008775.1 XP_016864264.1 10 2374
GenBank mRNA AB018320.1 - 6 695
AF049884.1 - 7 1060
AF049885.1 - 6 425
AK056628.1 - 9 881
AK225327.1 - 8 1220
AK225812.1 - 7 1306
AK293400.1 - 6 579
AK295742.1 - 7 614
BC011883.1 - 7 749 3677070

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin filament organization
cell adhesion
biological_process
cell migration
cell growth involved in cardiac muscle cell development
structural constituent of cytoskeleton
protein binding
cytoskeletal adaptor activity
structural constituent of muscle
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0