This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRTAP8-1 SIRNA Assays ›
Gene Symbol:
KRTAP8-1
Gene Name:
keratin associated protein 8-1
Gene Aliases:
KAP8.1
Chromosome Location:
Chr.21: 30812697 - 30813252 on Build GRCh38
UniGene ID:
Hs.407655
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 44096

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTAP8-1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175857.3 NP_787053.1 1 204
GenBank mRNA AB096961.1 - 1 202
AJ457064.1 - 1 204
BC130397.1 - 1 195 40146428
BC133033.1 - 1 195 40147055

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0