This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HUWE1 SIRNA Assays ›
Gene Symbol:
HUWE1
Gene Name:
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases:
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location:
Chr.X: 53532096 - 53686729 on Build GRCh38
UniGene ID:
Hs.136905
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
43780
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HUWE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031407.6
NP_113584.3
29
3552
XM_005261965.3
XP_005262022.1
28
3619
XM_011530747.1
XP_011529049.1
30
3795
XM_011530751.2
XP_011529053.1
29
3663
XM_017029191.1
XP_016884680.1
28
6547
XM_017029192.1
XP_016884681.1
28
6547
XM_017029193.1
XP_016884682.1
28
6526
XM_017029194.1
XP_016884683.1
28
6547
XM_017029195.1
XP_016884684.1
28
6547
XM_017029196.1
XP_016884685.1
28
6547
XM_017029197.1
XP_016884686.1
28
6547
XM_017029198.1
XP_016884687.1
29
3889
XM_017029199.1
XP_016884688.1
30
3816
XM_017029200.1
XP_016884689.1
29
3686
XM_017029201.1
XP_016884690.1
29
3703
XM_017029202.1
XP_016884691.1
29
3679
XM_017029203.1
XP_016884692.1
29
3685
XM_017029204.1
XP_016884693.1
27
6297
XM_017029205.1
XP_016884694.1
27
6277
XM_017029206.1
XP_016884695.1
28
6547
GenBank mRNA
AB071605.1
-
2
251
AK056671.1
-
12
1778
AK091352.1
-
1
70
AK294575.1
-
16
2152
AY772009.1
-
27
3197
AY929612.1
-
26
3149
DQ097177.1
-
29
3551
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
base-excision repair
protein monoubiquitination
histone ubiquitination
cell differentiation
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
DNA binding
ubiquitin-protein transferase activity
protein binding
ligase activity
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0