This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MRPL43 SIRNA Assays ›
Gene Symbol:
MRPL43
Gene Name:
mitochondrial ribosomal protein L43
Gene Aliases:
L43mt, MRP-L43, bMRP36a
Chromosome Location:
Chr.10: 100977822 - 100987519 on Build GRCh38
UniGene ID:
Hs.421848
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 43233

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MRPL43
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308396.1 NP_001295325.1 2, 3 307
NM_032112.2 NP_115488.2 2, 3 303
NM_176792.2 NP_789762.1 2, 3 303
NM_176793.1 NP_789763.1 2, 3 303
NM_176794.1 NP_789764.1 2, 3 303
XM_005270231.2 XP_005270288.1 2, 3 298
XM_006718035.3 XP_006718098.1 2, 3 298
GenBank mRNA AB049656.1 - 2, 3 266
AK095556.1 - 2, 3 287
AK291069.1 - 2, 3 258
AK300237.1 - 2, 3 285
BC015905.1 - 2, 3 303 3921157
BC041165.1 - 2, 3 237 5590251
BC052639.1 - 2, 3 244 6501303
BC053373.1 - 2, 3 256 6020112
BU597385.1 - 2, 3 308

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

translation
mitochondrial translational elongation
mitochondrial translational termination
structural constituent of ribosome
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0