This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZDHHC22 SIRNA Assays ›
Gene Symbol:
ZDHHC22
Gene Name:
zinc finger DHHC-type containing 22
Gene Aliases:
C14orf59
Chromosome Location:
Chr.14: 77131270 - 77146265 on Build GRCh38
UniGene ID:
Hs.525485
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 41576

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZDHHC22
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174976.2 NP_777636.2 3 1785
XM_006720119.3 XP_006720182.1 3 2015
XM_011536661.2 XP_011534963.1 3 5654
GenBank mRNA AK095612.1 - 3 1501
BC035881.1 - 2 1164
BC117676.2 - 2 1610 40069732

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein palmitoylation
protein localization to plasma membrane
protein binding
zinc ion binding
protein-cysteine S-palmitoyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0