This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CCDC107 SIRNA Assays ›
Gene Symbol:
CCDC107
Gene Name:
coiled-coil domain containing 107
Gene Aliases:
PSEC0222
Chromosome Location:
Chr.9: 35658282 - 35661503 on Build GRCh38
UniGene ID:
Hs.745107
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 41480

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CCDC107
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195217.1 NP_001182146.1 5 580
NM_174923.2 NP_777583.2 5 608
XM_005251403.4 XP_005251460.1 5 591
GenBank mRNA AK075523.1 - 5 566
BC018758.1 - 5 584 4907976

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header