This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF610 SIRNA Assays ›
Gene Symbol:
ZNF610
Gene Name:
zinc finger protein 610
Gene Aliases:
-
Chromosome Location:
Chr.19: 52336245 - 52367123 on Build GRCh38
UniGene ID:
Hs.147025
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 41008

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF610
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161425.1 NP_001154897.1 6 1832
NM_001161426.1 NP_001154898.1 6 1719
NM_001161427.1 NP_001154899.1 5 1703
NM_173530.2 NP_775801.2 6 1688
GenBank mRNA AK093359.1 - 6 1688
AK290888.1 - 6 1769
AL831871.1 - 1 576
BC039903.2 - 5 1647 6049284
BC105967.1 - 6 1606 6154656
DB577508.1 - 1 47

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0