This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C11ORF31 SIRNA Assays ›
Gene Symbol:
C11orf31
Gene Name:
chromosome 11 open reading frame 31
Gene Aliases:
C17orf10, SELH
Chromosome Location:
Chr.11: 57741250 - 57743554 on Build GRCh38
UniGene ID:
Hs.655373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 40334

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C11orf31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321335.1 NP_001308264.1 3 690
NM_170746.3 NP_734467.1 3 690
GenBank mRNA AF536829.1 - 3 586
BC021122.1 - 3 404 4766360
BG714263.1 - 3 405

Back To Top

More Information


Gene Ontology Categories:

Process(es)

poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0