This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FBXW8 SIRNA Assays ›
Gene Symbol:
FBXW8
Gene Name:
F-box and WD repeat domain containing 8
Gene Aliases:
FBW6, FBW8, FBX29, FBXO29, FBXW6
Chromosome Location:
Chr.12: 116910949 - 117032498 on Build GRCh38
UniGene ID:
Hs.624537
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 40142

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FBXW8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012174.1 NP_036306.1 5 689
NM_153348.2 NP_699179.2 5 887
XM_005253867.2 XP_005253924.1 4 745
XM_005253868.3 XP_005253925.1 4 578
XM_017019175.1 XP_016874664.1 5 664
XM_017019176.1 XP_016874665.1 6 896
XM_017019177.1 XP_016874666.1 6 896
GenBank mRNA AF176707.1 - 5 667
AK289621.1 - 5 680
BC037296.2 - 5 887 4821240

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Golgi organization
cell proliferation
protein ubiquitination
positive regulation of dendrite morphogenesis
spongiotrophoblast layer development
labyrinthine layer blood vessel development
ubiquitin-protein transferase activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header