This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SNX33 SIRNA Assays ›
Gene Symbol:
SNX33
Gene Name:
sorting nexin 33
Gene Aliases:
SH3PX3, SH3PXD3C, SNX30
Chromosome Location:
Chr.15: 75649007 - 75658627 on Build GRCh38
UniGene ID:
Hs.8705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 40120

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNX33
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318146.1 NP_001305075.1 1 347
NM_153271.1 NP_695003.1 1 347
GenBank mRNA AL833039.1 - 1 311
BC018775.1 - 1 347 4869639
EF219141.1 - 1 272
EF653821.1 - 1 513

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

mitotic cytokinesis
intracellular protein transport
endocytosis
endosome organization
mitotic nuclear division
vesicle organization
endosomal transport
protein import
cleavage furrow formation
macropinocytosis
negative regulation of endocytosis
positive regulation of membrane protein ectodomain proteolysis
membrane tubulation
negative regulation of protein localization to cell surface
positive regulation of protein localization to cell surface
protein binding
phosphatidylinositol binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0