This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM139 SIRNA Assays ›
Gene Symbol:
TMEM139
Gene Name:
transmembrane protein 139
Gene Aliases:
-
Chromosome Location:
Chr.7: 143284899 - 143288049 on Build GRCh38
UniGene ID:
Hs.17558
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 40047

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM139
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242773.2 NP_001229702.1 4 571
NM_001242774.2 NP_001229703.1 5 682
NM_001242775.2 NP_001229704.1 4 595
NM_001282876.1 NP_001269805.1 3 893
NM_001282877.1 NP_001269806.1 4 574
NM_153345.3 NP_699176.1 3 473
NR_040003.2 2 631
NR_104250.1 3 312
NR_104251.1 2 211
NR_104252.1 3 309
NR_104253.1 2 235
NR_104254.1 3 333
GenBank mRNA AK075067.1 - 3 418
AK172816.1 - 5 624
AK315170.1 - 4 519
AY358781.1 - 3 488
BC035517.1 - 3 873 5207904
BC035888.1 - 2 234
BE018738.1 - 2 528
BE562464.1 - 2 218
BF036500.1 - 1 131
BI761527.1 - 2 161
CB142336.1 - 3 260
CB156055.1 - 2 151
DA626625.1 - 4 534
DB222115.1 - 3 252
DT220400.1 - 2 163
R69052.1 - 3 258

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0