This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other PLD5 SIRNA Assays ›
Gene Symbol:
PLD5
Gene Name:
phospholipase D family member 5
Gene Aliases:
PLDC
Chromosome Location:
Chr.1: 242085051 - 242530546 on Build GRCh38
UniGene ID:
Hs.672452
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39617

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: PLD5
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195811.1 NP_001182740.1 4 530
NM_001195812.1 NP_001182741.1 3 368
NM_001320272.1 NP_001307201.1 5 824
NM_152666.2 NP_689879.2 5 817
XM_006711752.2 XP_006711815.1 4 984
XM_011544115.2 XP_011542417.1 6 892
XM_011544116.2 XP_011542418.1 6 720
XM_017000567.1 XP_016856056.1 4 997
XM_017000568.1 XP_016856057.1 4 518
GenBank mRNA AK091691.1 - 3 342
AK098092.1 - 5 824
AK295421.1 - 4 511
AK307857.1 - 4 509
AY391836.1 - 3 380
AY461578.1 - 4 530
BC101373.1 - 4 457 40025860
BC101374.1 - 4 457 40025861
BC101375.1 - 4 457 40025862

Back To Top

More Information


Gene Ontology Categories:

Process(es)

catalytic activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0