This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF182 SIRNA Assays ›
Gene Symbol:
RNF182
Gene Name:
ring finger protein 182
Gene Aliases:
-
Chromosome Location:
Chr.6: 13924446 - 13980009 on Build GRCh38
UniGene ID:
Hs.111164
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39579

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF182
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165032.1 NP_001158504.1 4 792
NM_001165033.1 NP_001158505.1 2 571
NM_001165034.1 NP_001158506.1 3 679
NM_152737.3 NP_689950.1 3 709
XM_017010448.1 XP_016865937.1 4 729
XM_017010449.1 XP_016865938.1 3 793
XM_017010450.1 XP_016865939.1 2 777
GenBank mRNA AK090576.1 - 2 528
AK315528.1 - 3 690
BC030666.1 - 3 609 5259219
BC050030.1 - 2 433 5297801

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
ubiquitin-protein transferase activity
protein binding
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0