This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SYNE3 SIRNA Assays ›
Gene Symbol:
SYNE3
Gene Name:
spectrin repeat containing nuclear envelope family member 3
Gene Aliases:
C14orf49, NET53, Nesp3
Chromosome Location:
Chr.14: 95416083 - 95516693 on Build GRCh38
UniGene ID:
Hs.354645
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39366

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SYNE3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152592.3 NP_689805.3 14 2424
XM_005267377.3 XP_005267434.1 15 2521
XM_006720063.3 XP_006720126.1 15 2621
XM_011536515.2 XP_011534817.1 15 2556
XM_017021056.1 XP_016876545.1 14 2408
GenBank mRNA AK098471.1 - 9 1187
AK131436.1 - 15 2274
BC146604.1 - 15 2421 40123293

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cytoskeleton organization
regulation of cell shape
establishment of protein localization to membrane
cytoskeletal anchoring at nuclear membrane
protein binding
actin filament binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm