This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C3ORF30 SIRNA Assays ›
Gene Symbol:
C3orf30
Gene Name:
chromosome 3 open reading frame 30
Gene Aliases:
TSCPA
Chromosome Location:
Chr.3: 119146150 - 119151455 on Build GRCh38
UniGene ID:
Hs.730099
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39135

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C3orf30
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152539.2 NP_689752.2 2 1508
GenBank mRNA AK057421.1 - 2 1508
BC130475.1 - 2 1507 40146506
BC144197.1 - 2 1507 9052717

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm