This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF785 SIRNA Assays ›
Gene Symbol:
ZNF785
Gene Name:
zinc finger protein 785
Gene Aliases:
-
Chromosome Location:
Chr.16: 30573219 - 30586038 on Build GRCh38
UniGene ID:
Hs.513509
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 39065

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF785
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152458.6 NP_689671.2 3 533
XM_011545752.2 XP_011544054.1 3 839
XM_011545753.2 XP_011544055.1 3 839
XM_017022967.1 XP_016878456.1 3 800
GenBank mRNA AK056692.1 - 3 489
AK292465.1 - 3 417
AK298848.1 - 3 512
AK309511.1 - 3 290
BC040642.2 - 3 451 4811661
CN401082.1 - 3 533

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0