This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM199 SIRNA Assays ›
Gene Symbol:
TMEM199
Gene Name:
transmembrane protein 199
Gene Aliases:
C17orf32, CDG2P, VMA12, VPH2
Chromosome Location:
Chr.17: 28357581 - 28363683 on Build GRCh38
UniGene ID:
Hs.594696
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 38978

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM199
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152464.2 NP_689677.1 2 342
GenBank mRNA AK223584.1 - 2 276
AK291475.1 - 2 265
AK311363.1 - 224
AY074907.1 - 2 252
BC033113.1 - 2 259 5429060
DC327155.1 - 2 342

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header