This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C19ORF25 SIRNA Assays ›
Gene Symbol:
C19orf25
Gene Name:
chromosome 19 open reading frame 25
Gene Aliases:
-
Chromosome Location:
Chr.19: 1473201 - 1479552 on Build GRCh38
UniGene ID:
Hs.532840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 38901

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C19orf25
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152482.2 NP_689695.2 2 176
XM_005259506.3 XP_005259563.1 2 192
XM_006722653.3 XP_006722716.1 2 151
XM_017026374.1 XP_016881863.1 2 499
GenBank mRNA AK075267.1 - 2 159
AK093985.1 - 2 141
AK297944.1 - 2 163
AL122089.1 - 2 147
AY007120.1 - 1 112
BC018441.1 - 2 146 4471390
BP210473.1 - 2 176

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header