This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LONRF1 SIRNA Assays ›
Gene Symbol:
LONRF1
Gene Name:
LON peptidase N-terminal domain and ring finger 1
Gene Aliases:
RNF191
Chromosome Location:
Chr.8: 12721897 - 12755485 on Build GRCh38
UniGene ID:
Hs.180178
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 38362

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LONRF1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152271.3 NP_689484.3 8 1654
XM_005273685.3 XP_005273742.1 8 1623
XM_011544694.2 XP_011542996.1 8 1406
GenBank mRNA AK074329.1 - 3 206
AK297266.1 - 7 791
AK300538.1 - 4 400
BC106042.1 - 8 920
BC117381.1 - 5 585 40125932
BC117385.1 - 5 585 40125936
BQ718281.1 - 251

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
proteolysis
ATP-dependent peptidase activity
ubiquitin-protein transferase activity
protein binding
zinc ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0