This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other LUZP6 SIRNA Assays ›
Gene Symbol:
LUZP6 MTPN
Gene Name:
leucine zipper protein 6
myotrophin
Gene Aliases:
MPD6, MTPNUT
GCDP, V-1
Chromosome Location:
Chr.7: 135926755 - 135977456 on Build GRCh38
Chromosome Location:
Chr.7: 135926755 - 135977456 on Build GRCh38
UniGene ID:
Hs.602015
Hs.602015
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 38305

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: LUZP6
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128619.2 NP_001122091.2 4 3073
Gene Symbol: MTPN
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145808.3 NP_665807.1 4 3073
GenBank mRNA AK055660.1 - 1 1624
AL110137.1 - 1 452
AL834231.1 - 4 2945
AY611627.1 - 1 271

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of translation
catecholamine metabolic process
positive regulation of macromolecule biosynthetic process
positive regulation of cardiac muscle hypertrophy
cell growth
regulation of striated muscle tissue development
cerebellar granule cell differentiation
neuron differentiation
positive regulation of cell growth
skeletal muscle tissue regeneration
positive regulation of NF-kappaB transcription factor activity
striated muscle cell differentiation
positive regulation of protein metabolic process
cellular response to mechanical stimulus
regulation of barbed-end actin filament capping
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0