This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other EVI5L SIRNA Assays ›
Gene Symbol:
EVI5L
Gene Name:
ecotropic viral integration site 5 like
Gene Aliases:
-
Chromosome Location:
Chr.19: 7830275 - 7864976 on Build GRCh38
UniGene ID:
Hs.26870
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 38046

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: EVI5L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159944.2 NP_001153416.1 19 3705
NM_145245.4 NP_660288.1 19 3783
XM_005272458.4 XP_005272515.1 20 3810
XM_017026239.1 XP_016881728.1 20 3802
GenBank mRNA BC014111.2 - 19 3701 4542709

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
regulation of vesicle fusion
activation of GTPase activity
negative regulation of cilium assembly
GTPase activator activity
protein binding
Rab GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0