This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other IMMP1L SIRNA Assays ›
Gene Symbol:
IMMP1L
Gene Name:
inner mitochondrial membrane peptidase subunit 1
Gene Aliases:
IMMP1, IMP1, IMP1-LIKE
Chromosome Location:
Chr.11: 31432399 - 31509644 on Build GRCh38
UniGene ID:
Hs.502223
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 37582

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: IMMP1L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304274.1 NP_001291203.1 4 415
NM_144981.2 NP_659418.1 5 478
XM_005252812.3 XP_005252869.1 6 599
XM_011519942.1 XP_011518244.1 6 536
XM_011519943.2 XP_011518245.1 7 657
XM_011519944.2 XP_011518246.1 6 565
XM_011519945.2 XP_011518247.1 5 480
XM_011519946.2 XP_011518248.1 6 525
XM_011519947.2 XP_011518249.1 5 462
XM_017017305.1 XP_016872794.1 5 536
XM_017017306.1 XP_016872795.1 4 378
XM_017017307.1 XP_016872796.1 4 392
GenBank mRNA AK057788.1 - 5 472
AK310496.1 - 4 395
AK311611.1 - 2 127
BC023595.2 - 4 361 4841844
DB288287.1 - 6 537

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

signal peptide processing
protein processing involved in protein targeting to mitochondrion
biological_process
mitochondrial respiratory chain complex assembly
molecular_function
serine-type endopeptidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0