This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SPATA4 SIRNA Assays ›
Gene Symbol:
SPATA4
Gene Name:
spermatogenesis associated 4
Gene Aliases:
SPEF1B, TSARG2
Chromosome Location:
Chr.4: 176184574 - 176195671 on Build GRCh38
UniGene ID:
Hs.481235
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 37307

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SPATA4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144644.3 NP_653245.2 1 221
GenBank mRNA AY040204.3 - 1 221
BC021731.2 - 1 122 5269042
HY030172.1 - 1 182

Back To Top

More Information


Gene Ontology Categories:

Process(es)

molecular_function
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header