This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM125 SIRNA Assays ›
Gene Symbol:
TMEM125
Gene Name:
transmembrane protein 125
Gene Aliases:
-
Chromosome Location:
Chr.1: 43269921 - 43274002 on Build GRCh38
UniGene ID:
Hs.104476
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 37294

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM125
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320244.1 NP_001307173.1 4 1327
NM_144626.2 NP_653227.1 4 1232
XM_005270467.3 XP_005270524.1 3 1472
XM_011540705.2 XP_011539007.1 4 1458
GenBank mRNA BC016858.1 - 3 1085 3845811
BC072393.1 - 4 1315

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0