This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TMEM129 SIRNA Assays ›
Gene Symbol:
TMEM129
Gene Name:
transmembrane protein 129
Gene Aliases:
D4S2561E
Chromosome Location:
Chr.4: 1715952 - 1721357 on Build GRCh38
UniGene ID:
Hs.518562
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 36687

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM129
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127266.1 NP_001120738.1 4 1768
NM_138385.3 NP_612394.1 3 1608
XM_005248038.4 XP_005248095.1 4 1640
XM_005248039.4 XP_005248096.1 4 1209
XM_011513594.2 XP_011511896.1 4 1218
XM_017008834.1 XP_016864323.1 4 1505
GenBank mRNA AK098466.1 - 3 623
AK310320.1 - 3 680
BC009331.2 - 3 1608
BC018921.2 - 3 1608
BC126370.1 - 5 1658 8992086

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
biological_process
ER-associated ubiquitin-dependent protein catabolic process
retrograde protein transport, ER to cytosol
molecular_function
ligase activity
metal ion binding
ubiquitin protein ligase activity involved in ERAD pathway

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0