This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SLC35A4 SIRNA Assays ›
Gene Symbol:
SLC35A4
Gene Name:
solute carrier family 35 member A4
Gene Aliases:
-
Chromosome Location:
Chr.5: 140564565 - 140569104 on Build GRCh38
UniGene ID:
Hs.406840
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 36057

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SLC35A4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080670.3 NP_542401.1 3 1604
GenBank mRNA AF445025.1 - 2 2857
AJ420598.1 - 1 617
AK289378.1 - 1 958
AL833972.1 - 2 1323
BC009906.2 - 3 1334 2961449

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

carbohydrate transport
proton transport
sugar:proton symporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0