This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KLHL4 SIRNA Assays ›
Gene Symbol:
KLHL4
Gene Name:
kelch like family member 4
Gene Aliases:
DKELCHL, KHL4
Chromosome Location:
Chr.X: 87517541 - 87670050 on Build GRCh38
UniGene ID:
Hs.49075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 35697

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KLHL4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019117.4 NP_061990.2 1 272
NM_057162.2 NP_476503.1 1 272
GenBank mRNA AB051474.1 - 1 218
AF284765.1 - 1 179
AF284766.1 - 1 179
AK056457.1 - 1 240
AL832514.1 - 1 212
BC140839.1 - 1 141 9021707
BC140840.1 - 1 141 9021708
BC142652.1 - 1 118 40148044
BC146677.1 - 1 118 40148894
BC161675.1 - 1 218 8862604
DB083610.1 - 1 272

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein ubiquitination
molecular_function
actin binding
ubiquitin-protein transferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0