This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF41 SIRNA Assays ›
Gene Symbol:
ZNF41
Gene Name:
zinc finger protein 41
Gene Aliases:
MRX89
Chromosome Location:
Chr.X: 47444691 - 47483671 on Build GRCh38
UniGene ID:
Hs.496074
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 3503

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF41
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324139.1 NP_001311068.1 4 382
NM_001324140.1 NP_001311069.1 5 672
NM_001324141.1 NP_001311070.1 4 411
NM_001324142.1 NP_001311071.1 5 688
NM_001324143.1 NP_001311072.1 4 724
NM_001324144.1 NP_001311073.1 5 762
NM_001324145.1 NP_001311074.1 4 427
NM_001324147.1 NP_001311076.1 5 733
NM_001324148.1 NP_001311077.1 5 778
NM_001324149.1 NP_001311078.1 4 451
NM_001324150.1 NP_001311079.1 6 860
NM_001324151.1 NP_001311080.1 5 802
NM_001324152.1 NP_001311081.1 5 848
NM_001324153.1 NP_001311082.1 5 712
NM_001324154.1 NP_001311083.1 4 599
NM_001324155.1 NP_001311084.1 4 623
NM_007130.3 NP_009061.1 5 1075
NM_153380.3 NP_700359.1 5 948
XM_006724550.3 XP_006724613.1 5 801
XM_006724555.3 XP_006724618.1 5 1096
XM_017029810.1 XP_016885299.1 5 1176
XM_017029811.1 XP_016885300.1 5 1237
XM_017029812.1 XP_016885301.1 5 1158
XM_017029813.1 XP_016885302.1 5 707
XM_017029814.1 XP_016885303.1 5 1147
XM_017029815.1 XP_016885304.1 5 1196
XM_017029816.1 XP_016885305.1 5 635
XM_017029817.1 XP_016885306.1 5 595
GenBank mRNA AB209520.1 - 1 784
AJ010017.1 - 4 391
AJ010018.1 - 4 415
AJ010019.1 - 4 461
AJ010020.1 - 4 477
AJ010021.1 - 4 501
AK290021.1 - 5 729
AK294858.1 - 4 324
BC015023.1 - 5 958 3908304
BQ053634.1 - 5 616
DA567652.1 - 4 321
DA783869.1 - 4 382
X60155.2 - 5 714

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0