This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF382 SIRNA Assays ›
Gene Symbol:
ZNF382
Gene Name:
zinc finger protein 382
Gene Aliases:
KS1
Chromosome Location:
Chr.19: 36605305 - 36628597 on Build GRCh38
UniGene ID:
Hs.631591
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 35000

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF382
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256838.1 NP_001243767.1 5 2145
NM_032825.4 NP_116214.2 5 2170
GenBank mRNA AF513816.1 - 5 2150
AK027592.1 - 5 2150
AL832265.1 - 3 3534
AY227449.1 - 5 2150
BC038358.1 - 4 2607
BC132675.1 - 4 2115 40146697

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm