This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other TTYH2 SIRNA Assays ›
Gene Symbol:
TTYH2
Gene Name:
tweety family member 2
Gene Aliases:
C17orf29
Chromosome Location:
Chr.17: 74213557 - 74262020 on Build GRCh38
UniGene ID:
Hs.27935
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 34362

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TTYH2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032646.5 NP_116035.5 10, 11 1144
NM_052869.1 NP_443101.1 4, 5 954
XM_005257824.3 XP_005257881.1 10, 11 1072
GenBank mRNA AF319952.1 - 10, 11 1123
AK091085.1 - 8, 9 825
AK126955.1 - 4, 5 1362
AK296511.1 - 10, 11 1067
AK315163.1 - 10, 11 1130
BC005168.1 - 10, 11 1119
BC107492.1 - 10, 11 1140 40033786
BC107602.1 - 10, 11 1140 40033791
BX647912.1 - 4, 5 954

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

chloride transmembrane transport
chloride channel activity
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0