This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other FSD1L SIRNA Assays ›
Gene Symbol:
FSD1L
Gene Name:
fibronectin type III and SPRY domain containing 1 like
Gene Aliases:
CCDC10, CSDUFD1, FSD1CL, FSD1NL, MIR1
Chromosome Location:
Chr.9: 105442183 - 105552433 on Build GRCh38
UniGene ID:
Hs.136901
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 33454

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FSD1L
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145313.2 NP_001138785.1 3, 4 394
NM_001287191.1 NP_001274120.1 2, 3 298
NM_001287192.1 NP_001274121.1 2, 3 298
NM_031919.4 NP_114125.1 2, 3 298
XM_005252254.2 XP_005252311.1 3, 4 627
XM_005252257.3 XP_005252314.1 2, 3 293
XM_005252260.3 XP_005252317.1 2, 3 283
XM_011519077.2 XP_011517379.1 4, 5 285
XM_011519078.2 XP_011517380.2 3, 4 292
XM_011519079.2 XP_011517381.2 3, 4 294
XM_017015182.1 XP_016870671.1 3, 4 627
XM_017015183.1 XP_016870672.1 2, 3 293
XM_017015185.1 XP_016870674.1 3, 4 300
XM_017015187.1 XP_016870676.1 2, 3 293
GenBank mRNA AF088031.1 - 2, 3 287
AF316830.1 - 2, 3 161
AK296433.1 - 2, 3 209
AK299350.1 - 3, 4 326
AK299491.1 - 2, 3 209
AK309817.1 - 3, 4 635
BP219536.1 - 3, 4 366
BP229804.1 - 2, 3 298
BP365251.1 - 2, 3 288
DN999402.1 - 2, 3 141

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7bff9b55f5-ncxmj:80/100.66.128.142:80.
git-commit: 5f36da1a8ad9bc9722227bfd86ebc827ae27d083
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0