This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other RNF170 SIRNA Assays ›
Gene Symbol:
RNF170
Gene Name:
ring finger protein 170
Gene Aliases:
ADSA, SNAX1
Chromosome Location:
Chr.8: 42849637 - 42897288 on Build GRCh38
UniGene ID:
Hs.491626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 33132

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RNF170
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160223.1 NP_001153695.1 7 1249
NM_001160225.1 NP_001153697.1 7 898
NM_030954.3 NP_112216.3 7 1019
NR_027668.1 6 1175
NR_027669.1 6 945
XM_006716404.2 XP_006716467.1 8 1006
XM_006716405.2 XP_006716468.1 5 765
XM_011544666.2 XP_011542968.1 7 803
XM_017013881.1 XP_016869370.1 8 946
XM_017013882.1 XP_016869371.1 6 791
GenBank mRNA AF209504.1 - 2 356
AK027748.1 - 7 935
AK090864.1 - 6 1108
AK095625.1 - 1 447
AK225086.1 - 7 899
AK225568.1 - 7 915
AL136620.1 - 7 903
BC013422.1 - 6 810
BC039461.1 - 7 1006
BC058289.1 - 7 882

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
protein binding
zinc ion binding
ligase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0