This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZSCAN31 SIRNA Assays ›
Gene Symbol:
ZSCAN31
Gene Name:
zinc finger and SCAN domain containing 31
Gene Aliases:
ZNF20-Lp, ZNF310P, ZNF323
Chromosome Location:
Chr.6: 28324737 - 28358202 on Build GRCh38
UniGene ID:
Hs.656413
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 33005

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZSCAN31
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135215.1 NP_001128687.1 8 1748
NM_001135216.1 NP_001128688.1 4 1244
NM_001243241.1 NP_001230170.1 4 1169
NM_001243242.1 NP_001230171.1 4 866
NM_001243243.1 NP_001230172.1 3 768
NM_001243244.1 NP_001230173.1 3 757
NM_030899.4 NP_112161.3 4 1233
NM_145909.2 NP_665916.1 8 1370
XM_005249295.1 XP_005249352.1 4 1095
XM_005249296.4 XP_005249353.1 5 5370
XM_011514807.2 XP_011513109.1 6 5420
XM_011514808.2 XP_011513110.1 4 5216
XM_011514809.1 XP_011513111.1 4 1080
XM_011514811.2 XP_011513113.1 4 1595
XM_011514812.2 XP_011513114.1 4 1121
XM_011514813.2 XP_011513115.1 4 1584
XM_017011196.1 XP_016866685.1 6 5508
GenBank mRNA AF513019.1 - 8 1370
AK027060.1 - 1 259
AK057702.1 - 8 1370
AK095261.1 - 8 1748
AK225382.1 - 1 262
AL833416.1 - 3 528
AL833617.1 - 4 1245
BC008490.1 - 4 1007 4248596
BC065241.1 - 3 538 6016003

Back To Top

More Information


Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0