This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other THOC7 SIRNA Assays ›
Gene Symbol:
THOC7
Gene Name:
THO complex 7
Gene Aliases:
NIF3L1BP1, fSAP24, hTREX30
Chromosome Location:
Chr.3: 63833870 - 63864478 on Build GRCh38
UniGene ID:
Hs.288151
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 32341

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: THOC7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001285387.2 NP_001272316.1 7 664
NM_001285404.1 NP_001272333.1 8 913
NM_025075.3 NP_079351.2 8 782
XM_006713339.3 XP_006713402.1 8 731
GenBank mRNA AF104914.1 - 7 630
AK027098.1 - 8 692
AK225110.1 - 8 670
BC020599.2 - 7 630
BC065012.1 - 8 652 6167514

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription elongation from RNA polymerase II promoter
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
viral mRNA export from host cell nucleus
nucleic acid binding
RNA binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0