This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MAPK13 SIRNA Assays ›
Gene Symbol:
MAPK13
Gene Name:
mitogen-activated protein kinase 13
Gene Aliases:
MAPK 13, MAPK-13, PRKM13, SAPK4, p38delta
Chromosome Location:
Chr.6: 36130484 - 36144524 on Build GRCh38
UniGene ID:
Hs.178695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 321

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MAPK13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002754.4 NP_002745.1 4 479
NR_072996.1 4 479
GenBank mRNA AB209586.1 - 3 1437
AB451221.1 - 5 380
AB451344.1 - 5 380
AF004709.1 - 4 428
AF092535.1 - 4 401
AF100546.1 - 4 382
AK222922.1 - 4 371
AK308018.1 - 4 460
BC000433.2 - 4 421 2819932
BC001641.2 - 4 421 2819932
BC004428.1 - 4 421 2819932
BC085196.1 - 4 449 6107307
BM921729.1 - 4 479
BT007221.1 - 4 380
CR536490.1 - 4 380
CR541687.1 - 4 380
DB473379.1 - 4 443
DR006449.1 - 4 483
KU178281.1 - 4 380
KU178282.1 - 4 380
U93232.1 - 4 387
Y10488.1 - 4 402

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
transcription, DNA-templated
regulation of transcription, DNA-templated
response to stress
response to osmotic stress
cell cycle
peptidyl-serine phosphorylation
positive regulation of interleukin-6 production
intracellular signal transduction
vascular endothelial growth factor receptor signaling pathway
positive regulation of inflammatory response
protein serine/threonine kinase activity
MAP kinase activity
protein binding
ATP binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0