This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other DOK3 SIRNA Assays ›
Gene Symbol:
DOK3
Gene Name:
docking protein 3
Gene Aliases:
DOKL
Chromosome Location:
Chr.5: 177501904 - 177510507 on Build GRCh38
UniGene ID:
Hs.720849
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 31832

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DOK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144875.1 NP_001138347.1 2 237
NM_001144876.1 NP_001138348.1 2 319
NM_001308235.1 NP_001295164.1 2 429
NM_001308236.1 NP_001295165.1 2 252
NM_024872.2 NP_079148.2 2 208
XM_005265985.3 XP_005266042.1 2 265
XM_005265987.1 XP_005266044.1 2 265
XM_011534654.2 XP_011532956.1 2 142
XM_011534655.2 XP_011532957.1 2 318
XM_011534656.2 XP_011532958.1 2 119
XM_011534657.2 XP_011532959.1 2 275
XM_017009883.1 XP_016865372.1 1 246
XM_017009884.1 XP_016865373.1 1 246
GenBank mRNA AK026223.1 - 2 203
AK097258.1 - 2 275
AK307300.1 - 2 275
AL583057.3 - 2 383
BC004564.2 - 2 196 4054381
BI489608.1 - 2 229
CF138017.1 - 2 360
DA674730.1 - 2 209
DA931137.1 - 2 319

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Ras protein signal transduction
insulin receptor binding
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0