This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other SHCBP1 SIRNA Assays ›
Gene Symbol:
SHCBP1
Gene Name:
SHC binding and spindle associated 1
Gene Aliases:
PAL
Chromosome Location:
Chr.16: 46578591 - 46624933 on Build GRCh38
UniGene ID:
Hs.123253
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 31724

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SHCBP1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001324319.1 NP_001311248.1 2 208
NM_024745.4 NP_079021.3 2 205
NR_136738.1 2 208
XM_011523335.2 XP_011521637.1 2 3739
GenBank mRNA AK055931.1 - 2 176
AK292164.1 - 2 172
AK310430.1 - 2 174
AK315706.1 - 2 174
BC030699.1 - 2 180 4829528
BP245197.1 - 2 205

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

fibroblast growth factor receptor signaling pathway
regulation of neural precursor cell proliferation
protein binding
SH2 domain binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0