This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASB13 SIRNA Assays ›
Gene Symbol:
ASB13
Gene Name:
ankyrin repeat and SOCS box containing 13
Gene Aliases:
-
Chromosome Location:
Chr.10: 5638857 - 5666595 on Build GRCh38
UniGene ID:
Hs.445899
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 31311

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASB13
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024701.3 NP_078977.2 5 625
NR_024581.1 4 474
NR_037164.1 6 706
GenBank mRNA AF403031.1 - 5 607
AK023196.1 - 5 608
AK091935.1 - 4 474
AK292071.1 - 5 611
BC012056.1 - 6 683 4552643
CR457302.1 - 5 581

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

protein ubiquitination
intracellular signal transduction
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0