This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ASB8 SIRNA Assays ›
Gene Symbol:
ASB8
Gene Name:
ankyrin repeat and SOCS box containing 8
Gene Aliases:
PP14212
Chromosome Location:
Chr.12: 48147788 - 48157595 on Build GRCh38
UniGene ID:
Hs.432699
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 30711

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASB8
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319296.1 NP_001306225.1 5 620
NM_001319297.1 NP_001306226.1 4 480
NM_001319298.1 NP_001306227.1 5 585
NM_001319299.1 NP_001306228.1 5 581
NM_001319300.1 NP_001306229.1 5 538
NM_001319301.1 NP_001306230.1 5 534
NM_024095.4 NP_077000.1 4 484
GenBank mRNA AF464877.1 - 4 376
AK024908.1 - 4 385
AK125596.1 - 1 1231
AK289957.1 - 4 451
AK297013.1 - 4 389
BC001321.1 - 4 391 3456049
BM837218.1 - 4 484
BP195070.1 - 5 534
BP210910.1 - 5 503
CR457336.1 - 3 314
DA071750.1 - 5 542
DA979010.1 - 5 502

Back To Top

More Information


Gene Ontology Categories:

Function(s)

protein ubiquitination
intracellular signal transduction

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header