This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other C17ORF75 SIRNA Assays ›
Gene Symbol:
C17orf75
Gene Name:
chromosome 17 open reading frame 75
Gene Aliases:
NJMU-R1, SRI2
Chromosome Location:
Chr.17: 32328441 - 32350047 on Build GRCh38
UniGene ID:
Hs.655257
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 30072

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C17orf75
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022344.3 NP_071739.2 10 1095
XM_005258022.3 XP_005258079.1 11 1113
XM_017024940.1 XP_016880429.1 10 964
XM_017024941.1 XP_016880430.1 10 962
GenBank mRNA AB062437.1 - 4 317
AF052093.1 - 10 1022
AF305686.1 - 10 1051
BC026017.2 - 10 1041 3352761
BC035715.1 - 10 1049 5588164
BC047325.1 - 10 1035 5493477
BC054035.1 - 10 1029 6143106

Back To Top

More Information


Gene Ontology Categories:

Process(es)

molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0