This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF688 SIRNA Assays ›
Gene Symbol:
ZNF688
Gene Name:
zinc finger protein 688
Gene Aliases:
-
Chromosome Location:
Chr.16: 30569677 - 30572407 on Build GRCh38
UniGene ID:
Hs.301463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 290098

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF688
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024683.1 NP_001019854.1 3 420
NM_145271.3 NP_660314.1 3 1108
XM_005255139.2 XP_005255196.1 3 337
XM_005255140.2 XP_005255197.1 3 957
GenBank mRNA AK122680.1 - 3 1436
BC018997.2 - 3 387 4336167
BM011208.1 - 3 383
KU178870.1 - 3 330
KU178871.1 - 2 640
KU178872.1 - 2 216

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0