This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KRTAP5-7 SIRNA Assays ›
Gene Symbol:
KRTAP5-7
Gene Name:
keratin associated protein 5-7
Gene Aliases:
KRTAP5-3, KRTAP5.7
Chromosome Location:
Chr.11: 71527267 - 71528164 on Build GRCh38
UniGene ID:
Hs.710675
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 289237

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: KRTAP5-7
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012503.1 NP_001012521.1 1 710
GenBank mRNA AB126076.1 - 1 710
AJ628243.1 - 1 296
BC136967.1 - 1 699 9020959
BC136968.1 - 1 699 9020960

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0