This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other MYLK SIRNA Assays ›
Gene Symbol:
MYLK
Gene Name:
myosin light chain kinase
Gene Aliases:
AAT7, KRP, MLCK, MLCK1, MLCK108, MLCK210, MSTP083, MYLK1, smMLCK
Chromosome Location:
Chr.3: 123612296 - 123884302 on Build GRCh38
UniGene ID:
Hs.477375
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 288626

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: MYLK
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321309.1 NP_001308238.1 33 6741
NM_053025.3 NP_444253.3 34 6949
NM_053026.3 NP_444254.3 33 6742
NM_053027.3 NP_444255.3 33 6796
NM_053028.3 NP_444256.3 32 6589
XM_017006469.1 XP_016861958.1 17 4225
XM_017006470.1 XP_016861959.1 16 3213
XM_017006471.1 XP_016861960.1 16 3266
XM_017006472.1 XP_016861961.1 4 1559
XM_017006473.1 XP_016861962.1 4 1508
GenBank mRNA AF172084.1 - 1 814
AK025953.1 - 1 721
AK127437.1 - 1 2471
BC015838.1 - 1 71
BC064420.1 - 3 1622
BC113456.1 - 3 1430
BC113458.1 - 3 1425
CA391924.1 - 1 350
CR749364.1 - 3 6408

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
protein phosphorylation
muscle contraction
smooth muscle contraction
tonic smooth muscle contraction
positive regulation of cell migration
bleb assembly
positive regulation of calcium ion transport
aorta smooth muscle tissue morphogenesis
cellular hypotonic response
positive regulation of wound healing
mitophagy in response to mitochondrial depolarization
xenophagy
actin binding
myosin light chain kinase activity
protein binding
calmodulin binding
ATP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0