siRNA Details
Gene Symbol: Olfr206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146991.1 | NP_667202.1 | 1 | 554 | ||
GenBank mRNA | BC138396.1 | - | 1 | 581 | 8861416 | |
BC138397.1 | - | 1 | 581 | 8861417 |

Gene Symbol: Olfr206 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146991.1 | NP_667202.1 | 1 | 554 | ||
GenBank mRNA | BC138396.1 | - | 1 | 581 | 8861416 | |
BC138397.1 | - | 1 | 581 | 8861417 |