This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other KCNIP2 SIRNA Assays ›
Gene Symbol:
Kcnip2 LOC100911951
Gene Name:
Kv channel-interacting protein 2
Kv channel-interacting protein 2-like
Gene Aliases:
Kchip2
-
Chromosome Location:
Chr.1: 265549610 - 265573397 on Build Rnor_6.0
Chromosome Location:
Chr.1: 262892549 - 262916345 on Build Rnor_6.0
UniGene ID:
Rn.98384
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 286766

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 261.00
Your Price
Online offer: 193.14
193.1
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Kcnip2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033961.1 NP_001029133.1 8 1053
NM_020094.2 NP_064479.2 10 1203
NM_020095.2 NP_064480.2 9 1149
XM_008760448.1 XP_008758670.1 8 2761
XM_008760449.1 XP_008758671.1 10 1312
XM_008760450.1 XP_008758672.1 8 953
GenBank mRNA BC085905.1 - 8 1053 7193750
Gene Symbol: LOC100911951
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006231581.2 XP_006231643.1 8 949
XM_008760479.1 XP_008758701.1 10 1260
XM_008760480.1 XP_008758702.1 9 1205
XM_008760481.1 XP_008758703.1 8 1267
XM_008760482.1 XP_008758704.1 8 1108
XM_008760483.1 XP_008758705.1 10 1312

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

clustering of voltage-gated potassium channels
protein heterooligomerization
potassium ion export
potassium ion transmembrane transport
membrane repolarization
regulation of potassium ion transmembrane transport
voltage-gated ion channel activity
A-type (transient outward) potassium channel activity
potassium channel activity
calcium ion binding
protein binding
potassium channel regulator activity
identical protein binding
ion channel binding
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0