This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other VOM1R27 SIRNA Assays ›
Gene Symbol:
Vom1r27 LOC100911644
Gene Name:
vomeronasal 1 receptor 27
vomeronasal type-1 receptor 4-like
Gene Aliases:
V1re19
-
Chromosome Location:
Chr.1: 68528976 - 68529893 on Build Rnor_6.0
Chromosome Location:
Chr.1: 68649663 - 68650566 on Build Rnor_6.0
UniGene ID:
Rn.129439
-
Species:
Rat
Species Specific ID (Flybase ID):
-
-
siRNA ID 286479

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Vom1r27
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008952.1 NP_001008952.1 1 676
GenBank mRNA AY510351.1 - 1 676
Gene Symbol: LOC100911644
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008758880.1 XP_008757102.1 1 664

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

G-protein coupled receptor signaling pathway
pheromone receptor activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header