This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other ZNF773 SIRNA Assays ›
Gene Symbol:
ZNF773
Gene Name:
zinc finger protein 773
Gene Aliases:
ZNF419B
Chromosome Location:
Chr.19: 57499854 - 57516423 on Build GRCh38
UniGene ID:
Hs.579576
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 285044

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ZNF773
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304334.1 NP_001291263.1 4 1512
NM_001304336.1 NP_001291265.1 4 1508
NM_198542.2 NP_940944.1 4 1515
GenBank mRNA AK057209.1 - 4 1431
BC005167.2 - 4 1428 3535599

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0