This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other CD177 SIRNA Assays ›
Gene Symbol:
CD177
Gene Name:
CD177 molecule
Gene Aliases:
HNA-2a, HNA2A, NB1, NB1 GP, PRV-1, PRV1
Chromosome Location:
Chr.19: 43353659 - 43366075 on Build GRCh38
UniGene ID:
Hs.232165
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 28486

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CD177
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020406.3 NP_065139.2 4, 5 569
XM_017027021.1 XP_016882510.1 4, 5 553
XM_017027022.1 XP_016882511.1 4, 5 553
GenBank mRNA AB237911.1 - 4, 5 521
AF146747.1 - 4, 5 548
AJ290452.1 - 4, 5 540
AJ305326.1 - 4, 5 659
AJ310433.1 - 4, 5 540
AK225226.1 - 4, 5 542
AK290965.1 - 4, 5 544
AY358932.1 - 4, 5 529
BC029167.1 - 4, 5 531 5182826
BT020111.1 - 4, 5 513
CD514938.1 - 4, 5 517
DA728784.1 - 4, 5 569

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
leukocyte migration
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0