This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
See other NT5M SIRNA Assays ›
Gene Symbol:
NT5M
Gene Name:
5',3'-nucleotidase, mitochondrial
Gene Aliases:
dNT-2, dNT2, mdN
Chromosome Location:
Chr.17: 17303366 - 17347663 on Build GRCh38
UniGene ID:
Hs.513977
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 28412

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NT5M
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020201.3 NP_064586.1 5 1484
XM_005256731.3 XP_005256788.1 6 1548
XM_005256732.3 XP_005256789.1 5 1491
XM_005256737.1 XP_005256794.1 6 1304
XM_011523963.1 XP_011522265.1 7 1341
XM_011523964.2 XP_011522266.1 7 1310
XM_011523965.2 XP_011522267.1 6 1214
XM_011523967.2 XP_011522269.1 6 1584
XM_017024879.1 XP_016880368.1 6 2244
GenBank mRNA AF210652.1 - 5 1515
BC035838.1 - 5 1458 5727827

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

DNA replication
pyrimidine deoxyribonucleotide catabolic process
dephosphorylation
dUMP catabolic process
pyrimidine nucleoside catabolic process
nucleotide binding
nucleotidase activity
5'-nucleotidase activity
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0